missing translation for 'onlineSavingsMsg'
Learn More

Filaggrin Antibody (rFLG/1945), DyLight 550, Novus Biologicals™

Product Code. 18392922
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392922 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392922 Supplier Bio-Techne Supplier No. NBP314280R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filaggrin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rFLG/1945
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Host Species Mouse
Immunogen Recombinant human Filaggrin protein fragment (around aa 198-288) (exact sequence is proprietary) (Uniprot: P20930 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.