missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Antibody (rFLG/1561), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08382APC
This item is not returnable.
View return policy
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Filaggrin | |
| Monoclonal | |
| APC | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary) (Uniprot: P20930) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 2312 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rFLG/1561 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction