missing translation for 'onlineSavingsMsg'
Learn More

Filaggrin Antibody (FLG/3909R), DyLight 680, Novus Biologicals™

Product Code. 18322837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322837 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322837 Supplier Novus Biologicals Supplier No. NBP314281FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filaggrin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone FLG/3909R
Conjugate DyLight 680
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Host Species Rabbit
Immunogen Recombinant human Filaggrin protein fragment (around aa 198-288) (exact sequence is proprietary) (Uniprot: P20930 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.