missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Filaggrin |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | FLG/3909R |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ATOD2, epidermal filaggrin, filaggrin, Profilaggrin |
| Host Species | Rabbit |
| Immunogen | Recombinant human Filaggrin protein fragment (around aa 198-288) (exact sequence is proprietary) (Uniprot: P20930 ) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?