missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Antibody (FLG/1561), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54366AF488
This item is not returnable.
View return policy
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Filaggrin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary) (Uniprot: P20930) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 2312 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| FLG/1561 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction