missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FIIN 1 hydrochloride

Product Code. 18005452
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 18005452

Brand: Tocris Bioscience™ 4002/2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, irreversible FGFR inhibitor

FIIN 1 hydrochloride is a potent, irreversible FGFR inhibitor (Kd values are 2.8, 5.4, 6.9 and 120 nM for FGFR1, FGFR3, FGFR2 and FGFR4 respectively); acts at the ATP binding site. Also irreversibly inhibits Flt-1, Flt-4 and VEGFR-2 (Kd values are 32, 120 and 210 nM respectively); displays limited activity (Kd >500 nM) at other kinases. Exhibits antiproliferative activity in FGFR3- and FGFR1-transformed Ba/F3 cells (EC50 values are 10 and 14 nM respectively). Derived from the reversible FGFR inhibitor, PD 173074 (Cat. No. 3044).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-((3-(2,6-dichloro-3,5-dimethoxyphenyl)-7-(4-(diethylamino)butylamino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)methyl)phenyl)acrylamide
CAS 1256152-35-8
Quantity 2 mg
Target FGF Receptor Inhibitors
Molecular Formula C32H39Cl2N7O4
Purity 96%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.