missing translation for 'onlineSavingsMsg'
Learn More

FIH-1/HIF-1AN Antibody, HRP, Novus Biologicals™

Product Code. 18333904 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333904 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333904 Supplier Novus Biologicals Supplier No. NBP299356H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FIH-1/HIF-1AN Polyclonal specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIH-1/HIF-1AN
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FIH-1/HIF-1AN.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55662
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.