missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIH-1/HIF-1AN Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99356APC
This item is not returnable.
View return policy
Description
FIH-1/HIF-1AN Polyclonal antibody specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| FIH-1/HIF-1AN | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FIH-1/HIF-1AN. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
| 55662 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction