missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIH-1/HIF-1AN Antibody (162c) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-30333JF635
This item is not returnable.
View return policy
Description
FIH-1/HIF-1AN Monoclonal antibody specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| FIH-1/HIF-1AN | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase | |
| Full length human Factor Inhibiting HIF-1 protein [Swiss-Prot# Q9NWT6] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
| 55662 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 162c | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction