missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17604-1-AP-20UL
This item is not returnable.
View return policy
Description
The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).Specifications
| FIGNL1 | |
| Polyclonal | |
| Unconjugated | |
| FIGNL1 | |
| fidgetin like 1, Fidgetin like protein 1, FIGNL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289777, 60530, 63979 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6GX84, Q6PIW4, Q8BPY9 | |
| Fignl1 | |
| FIGNL1 Fusion Protein Ag11709 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction