missing translation for 'onlineSavingsMsg'
Learn More

FICOLL™, White Powder, MP Biomedicals™

Product Code. 11460182 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
Product Code. Quantity unitSize
11460182 10 g 10g
11480182 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11460182

Brand: MP Biomedicals 0216000310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Density up to 1.2g/mL
Quantity 10 g
Formula Weight ≈400,000
Physical Form Solid
Chemical Name or Material FICOLL™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.