missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Description
Ficoll 400 (White to Off-White Powder/Molecular Biology)
Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
Specifications
Specifications
| Color | White |
| Physical Form | Powder/Solid |
| Quantity | 25 g |
| Formula Weight | ≈400,000 |
| Grade | Molecular Biology |
| Packaging | Poly Bottle |
| Specific Rotation | Inclusive between 50° (+ or -) |
| Chemical Name or Material | Ficoll™ 400 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?