missing translation for 'onlineSavingsMsg'
Learn More

Ficoll|r 400, Thermo Scientific Chemicals

Catalog Number 11459597
Click to view available options
Quantity:
10 g
50 g
26873-85-8
C15H27ClO12
434.819
MFCD00081599
IACFXVUNKCXYJM-AKSHDPDZSA-N
ficol 400
88330506
2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
26873-85-8
C15H27ClO12
434.819
MFCD00081599
IACFXVUNKCXYJM-AKSHDPDZSA-N
ficol 400
88330506
2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Ficoll™ 400 is used for cell separation and organelle isolation. It is also used to separate blood to its components like erythrocytes and leukocytes. It acts as a mononuclear cell separator and used in culture media, nucleic acidhybridization, electrophoresis and in immunological studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight ca 400,000
Chemical Name or Material Ficoll 400

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'