missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Ficoll
Description
Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles.
Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles. Also used in Denhardt's Reagent and in gel loading buffers.
Specifications
Specifications
| Chemical Name or Material | Ficoll |
| CAS | 26873-85-8 |
| Quantity | 25 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?