missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ficolin 3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£133.00 - £386.00
Specifications
| Antigen | Ficolin 3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17763384
|
Invitrogen™
PA5121988 |
20 μL |
£133.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17753384
![]() |
Invitrogen™
PA5121987 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.Specifications
| Ficolin 3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75636 | |
| 8547 | |
| Glu34-Met297 (Accession O75636), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FCN3 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Collagen/fibrinogen domain-containing lectin 3 p35; Collagen/fibrinogen domain-containing protein 3; FCN3; FCNH; ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen); ficolin 3; ficolin-3; HAKA1; Hakata antigen; LOW QUALITY PROTEIN: ficolin-3; MGC22543 | |
| FCN3 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title