missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ficolin 3 Monoclonal Antibody (296134)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524004
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human Ficolin-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
Specifications
| Ficolin 3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O75636 | |
| FCN3 | |
| Mouse myeloma cell line NS0-derived recombinant human Ficolin-3 Isoform 2 Lys22-Arg288. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 296134 | |
| Unconjugated | |
| FCN3 | |
| Collagen/fibrinogen domain-containing lectin 3 p35; Collagen/fibrinogen domain-containing protein 3; FCN3; FCNH; ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen); ficolin 3; ficolin-3; HAKA1; Hakata antigen; LOW QUALITY PROTEIN: ficolin-3; MGC22543 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8547 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction