missing translation for 'onlineSavingsMsg'
Learn More

Ficolin-3 Isoform 2 Antibody (296134) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30736615 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736615 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736615 Supplier Novus Biologicals Supplier No. FAB2367I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ficolin-3 Isoform 2 Monoclonal antibody specifically detects Ficolin-3 Isoform 2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ficolin-3 Isoform 2
Applications Western Blot
Classification Monoclonal
Clone 296134
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias FCN3, FCNH, HAKA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Ficolin-3, Lys22-Arg299, Accession # O75636-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.