missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-3 Antibody (FCN334), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41283C
This item is not returnable.
View return policy
Description
Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for ELISA.
Specifications
| Ficolin-3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FCN3 | |
| H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region. | |
| 0.1 mL | |
| 8547 | |
| Human | |
| Purified |
| ELISA | |
| FCN334 | |
| ELISA | |
| Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Reacts specifically with H-ficolin. No cross-reaction has been observed with L- or M-ficolin. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction