missing translation for 'onlineSavingsMsg'
Learn More

Ficolin-3 Antibody (FCN334), Biotin, Novus Biologicals™

Product Code. 18418731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418731 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418731 Supplier Novus Biologicals Supplier No. NBP223573B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ficolin-3 Monoclonal antibody specifically detects Ficolin-3 in Human samples. It is validated for ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ficolin-3
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone FCN334
Conjugate Biotin
Formulation PBS
Gene Alias Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin
Host Species Mouse
Immunogen Recombinant human H-ficolin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.