missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-3 Antibody (4H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62788
This item is not returnable.
View return policy
Description
Ficolin-3 Monoclonal specifically detects Ficolin-3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| Ficolin-3 | |
| Monoclonal | |
| 0.1mg/mL | |
| Immunohistochemistry, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunoassay | |
| Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
| 4H5 | |
| Unconjugated | |
| PBS and 0.1% bovine serum albumin, 0.02 NaN3 with 0.02% Sodium Azide | |
| FCN3 | |
| Ficolin-3 | |
| 50 μg | |
| 8547 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction