missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human, Rat samples. It is validated for Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay.
Specifications
Specifications
| Antigen | Ficolin-2 |
| Applications | Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay |
| Classification | Monoclonal |
| Clone | GN4 |
| Concentration | 0.1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay |
| Formulation | 0.2 um filtered solution in PBS, 0.1% BSA |
| Gene Alias | 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?