missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-2 Antibody (19), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05042CL1
This item is not returnable.
View return policy
Description
Ficolin-2 Monoclonal antibody specifically detects Ficolin-2 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Ficolin-2 | |
| Monoclonal | |
| CoraFluor 1 | |
| 37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35 | |
| Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| 19 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2220 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction