missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-1 Mouse anti-Human, Clone: 1, Novus Biologicals™
Mouse Monoclonal Antibody
£407.00
Specifications
| Antigen | Ficolin-1 |
|---|---|
| Clone | 1 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:20000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | ELISA |
Description
Ficolin-1 Monoclonal specifically detects Ficolin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Ficolin-1 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Collagen/fibrinogen domain-containing protein 1, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin-A, Ficolin-alpha, M-ficolin | |
| Purified prokariot expressed recombinant human M-Ficolin (aa 95 to 314) | |
| Primary | |
| Store at 4°C. Do not freeze. |
| 1 | |
| ELISA 1:20000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 2219 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title