missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-1 Antibody (2B7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002219-M06
This item is not returnable.
View return policy
Description
Ficolin-1 Monoclonal antibody specifically detects Ficolin-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Ficolin-1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Collagen/fibrinogen domain-containing protein 1, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin-A, Ficolin-alpha, M-ficolin | |
| FCN1 (NP_001994, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SSELRVDLVDFEGNHQFAKYKSFKVADEAEKYKLVLGAFVGGSAGNSLTGHNNNFFSTKDQDNDVSSSNCAEKFQGAWWYADCHASNLNGLYLMGPHESY | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2B7 | |
| Western Blot 1:500, ELISA | |
| NP_001994 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 2219 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction