missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-1 Antibody (1), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05050JF549
This item is not returnable.
View return policy
Description
Ficolin-1 Monoclonal specifically detects Ficolin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ficolin-1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FCN1 | |
| Purified prokariot expressed recombinant human M-Ficolin (aa 95 to 314) | |
| 0.1 mL | |
| 2219 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Collagen/fibrinogen domain-containing protein 1, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin-A, Ficolin-alpha, M-ficolin | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| M-Ficolin | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction