missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficodox™ Plus, Pure, Mixed COD Reagent, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Physical Form | Liquid |
| Melting Point | 3°C |
| Boiling Point | 280°C |
| Packaging | Plastic Coated Glass bottle |
| Color | Brown to Yellow |
| Quantity | 2.5 L |
Safety and Handling
Product Identifier
- Ficodox]TS Plus
- Danger
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 1 A
- H340-May cause genetic defects.
- H350-May cause cancer.
- H314-Causes severe skin burns and eye damage.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction