missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550497
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Fibulin 5 protein.
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications
| Fibulin 5 | |
| Polyclonal | |
| Unconjugated | |
| FBLN5 | |
| A55; ADCL2; ARCL1A; ARMD3; Dance; developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; Fbln5; FIBL-5; fibulin 5; fibulin-5; FLJ90059; HNARMD; testis tissue sperm-binding protein Li 75n; UNQ184/PRO210; UP50; urine p50 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10516 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 3.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UBX5 | |
| FBLN5 | |
| Synthetic peptide corresponding to a region derived from internal residues of human fibulin 5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction