missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 5 Monoclonal Antibody (3F8D5,7F8D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515293
This item is not returnable.
View return policy
Description
MA5-15293 targets Fibulin 5 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15293 immunogen is purified recombinant fragment of Fibulin 5 expressed in E. Coli.
Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications
| Fibulin 5 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q9UBX5 | |
| FBLN5 | |
| Purified recombinant fragment of Fibulin 5 expressed in E. Coli. | |
| RUO | |
| 10516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 3F8D5,7F8D5, 7F8D5 | |
| Unconjugated | |
| FBLN5 | |
| A55; ADCL2; ARCL1A; ARMD3; Dance; developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; Fbln5; FIBL-5; fibulin 5; fibulin-5; FLJ90059; HNARMD; testis tissue sperm-binding protein Li 75n; UNQ184/PRO210; UP50; urine p50 protein | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction