missing translation for 'onlineSavingsMsg'
Learn More

Fibulin 5/DANCE Antibody (293904) [DyLight 755], Novus Biologicals™

Product Code. 30734462 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734462 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734462 Supplier Novus Biologicals Supplier No. FAB3095Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibulin 5/DANCE Monoclonal antibody specifically detects Fibulin 5/DANCE in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 5/DANCE
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 293904
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, fibulin 5, UP50FLJ90059, Urine p50 protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fibulin 5/DANCE, Gln24-Phe448, Accession # Q9UBX5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10516
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.