Learn More
Invitrogen™ Fibulin 3 Recombinant Rabbit Monoclonal Antibody (K01_2M22)
Description
Immunogen sequence: QDTEETITYT QCTDGYEWDP VRQQCKDIDE CDIVPDACKG GMKCVNHYGG YLCLPKTAQI IVNNEQPQQE TQPAEGTSGA TTGVVAASSM ATSGVLPGGG FVASAAAVAG PEMQTGRNNF VIRRNPADPQ RIPSNPSHRI QCAAGYEQSE HNVCQDIDEC TAGTHNCRAD QVCINLRGSF ACQCPPGYQK RGEQCVDIDE CTIPPYCHQR CVNTPGSFYC QCSPGFQLAA NNYTCVDINE CDASNQCAQQ CYNILGSFIC QCNQGYELSS DRLNCEDIDE CRTSSYLCQY QCVNEPGKFS CMCPQGYQVV RSRTCQDINE CETTNECRED EMCWNYHGGF RCYPRNPCQD PYILTPENRC VCPVSNAMCR ELPQSIVYKY MSIRSDRSVP SDIFQIQATT IYANTINTFR IKSGNENGEF YLRQTSPVSA MLVLVKSLSG PREHIVDLEM LTVSSIGTFR TSSVLRLTII VGPFSF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%, Pig - 95%, Cynomolgus monkey - 99%.
Specifications
Specifications
| Antigen | Fibulin 3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2M22 |
| Concentration | 0.60 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | EFEMP1 |
| Gene Accession No. | Q12805 |
| Gene Alias | DHRD; DRAD; EFEMP1; EGF containing fibulin like extracellular matrix protein 1; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; extracellular protein S1-5; FBLN3; FBNL; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353; MLVT; MTLV; S1-5; T16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.