missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin-3/EFEMP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-691-0.025MG
This item is not returnable.
View return policy
Description
Fibulin-3/EFEMP1 Polyclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Fibulin-3/EFEMP1 | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 | |
| DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The Anti EFEMP1 selective was generated against a conserved sequence near the N terminal region that is unique to EFEMP1 protein only. This detects a strong 62 kDa EFEMP1 protein in human. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, ELISA | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| EFEMP1 | |
| Synthetic cyclic peptide corresponding to N terminal amino acids 25-42 of Human EFEMP1, peptide was covalently modified. Sequence: TYTQCTDGYEWDPVRQQC | |
| 0.025 mg | |
| Vision | |
| 2202 | |
| Rat, Monkey, Mouse, Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction