missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin-3/EFEMP1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77040AF350
This item is not returnable.
View return policy
Description
Fibulin-3/EFEMP1 Polyclonal antibody specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Fibulin-3/EFEMP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Fibulin 3. The immunogen is located within amino acids 410 - 460 of Fibulin 3. Amino Acid Squence: TFRIKSGNENGEFYLRQTS | |
| 0.1 mL | |
| Vision | |
| 2202 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction