missing translation for 'onlineSavingsMsg'
Learn More

Fibulin 1 Antibody (4C9), Novus Biologicals™

Product Code. 18326227 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326227 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326227 Supplier Novus Biologicals Supplier No. H00002192M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibulin 1 Monoclonal antibody specifically detects Fibulin 1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 4C9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22497
Gene Alias FBLN, FIBL1, FIBL-1, fibulin 1, fibulin-1
Host Species Mouse
Immunogen FBLN1 (AAH22497, 151 a.a. ~ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRDTGDEVVCSCFVGYQLLSDGVSCEDVNECITGSHSCRLGESCINTVGSFRCQRDSSCGT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2192
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.