missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibronectin Recombinant Rabbit Monoclonal Antibody (23GB1270), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Fibronectin |
|---|---|
| Clone | 23GB1270 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088919
![]() |
Invitrogen™
MA554174 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088282
|
Invitrogen™
MA554175 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Recombinant Monoclonal antibody specifically detects Fibronectin in Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Fibronectin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| P04937, P11276 | |
| 14268, 25661 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1270 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Anastellin; CIG; Cold insoluble globulin (CIG); Cold-insoluble globulin; cumulus cell-specific fibronectin 1 transcript; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; E330027I09; ED-B; embryo-specific fibronectin 1 transcript; ferritin L subunit; ferritin L-chain; FIBNEC; fibronectin; fibronectin 1; fibronectin ED-A- region; fibronectin ED-B+ region; fibronectin variable region; FINC; FN; FN1; fn-1; FNZ; GFND; GFND2; LETS; LOW QUALITY PROTEIN: fibronectin; Migration stimulating factor (MSF); migration-stimulating factor; MSF; Ugl-Y1; Ugl-Y2; Ugl-Y3 | |
| A synthesized peptide derived from human Fibronectin (1-2477AA). | |
| Primary | |
| -20°C | |
| FN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title