missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Rabbit anti-Human, Clone: 14, Novus Biologicals™
Rabbit Monoclonal Antibody
£306.00 - £439.00
Specifications
| Antigen | Fibronectin |
|---|---|
| Clone | 14 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18296896
|
Novus Biologicals
NBP2-89407-50UL |
50 μg |
£306.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18063616
|
Novus Biologicals
NBP2-89407-100UL |
100 μg |
£439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Fibronectin | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Recombinant Human Fibronectin protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 14 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 2335 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title