missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (HFN7.1), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34633PEATT594
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human, Mouse samples. It is validated for Flow Cytometry.
Spécification
| Fibronectin | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| FN1 | |
| Human fibronectin Purified from serum by affinity chromatography on gelatin-sepharose (Uniprot: P02751) | |
| 0.1 mL | |
| Primary | |
| Fibronectin is a dimeric glycoprotein of 440kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This monoclonal antibody is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to its receptor on the cell surface. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| HFN7.1 | |
| Flow Cytometry | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 2335.0 | |
| Human, Mouse | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu