missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fibronectin Antibody Blocking Peptide

Product Code. 18455322 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18455322

Brand: Novus Biologicals™ NBP191258PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocking Peptide

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2335
Concentration 1 mg/ml
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-91258. For further blocking peptide related protocol, click here .
Gene Alias CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Gene Symbol FN1
Quantity 0.1 mg
Research Category Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.