missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (2F4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80737
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Fibronectin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500 - 1:1000, Flow Cytometry 1:200 - 1:400, ELISA : 1:10000, Immunohistochemistry : 1:200 - 11:1000, Immunocytochemistry/Immunofluorescence : 1:100, Immunohistochemistry-Paraffin, CyTOF-ready | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 2F4 | |
| Unconjugated | |
| PBS | |
| FN1 | |
| Purified recombinant fragment of human Fibronectin expressed in E. coli. [UniProt# P02751] | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2335 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction