Learn More
STEMCELL Technologies Fibronectin
Product Code. 17149241
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
17149241 1 mg 1mg
1 options
Product Code. 17149241 Supplier STEMCELL Technologies Supplier No. 07159
For coating tissue cultureware to promote cell adherence
  • Fibronectin is used as a coating agent for coverslips and can also be used with DMEM/F-12 (Catalog #36254) and ITS Supplement-A (Catalog #07151) or ITS Supplement-B (Catalog #07155) to improve the yield of neural cells during the final stages of mouse embryonic stem cell in vitro differentiation into neural cell types
Specifications
Cell Type Pluripotent Stem Cells
Quantity 1 mg