missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibromodulin Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5149102

Product Code. 30535050

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fibromodulin
Polyclonal
Unconjugated
FMOD
AI131919; AU041740; Collagen-binding 59 kDa protein; fibromodulin; FM; FMOD; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin; SLRR2E
Sheep
Antigen affinity chromatography
RUO
14264, 2331, 64507
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
P50608, P50609, Q06828
FMOD
Mouse myeloma cell line NS0-derived recombinant human Fibromodulin/FMOD, Asp75-Ile376.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.