missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fibromodulin/FMOD Polyclonal antibody specifically detects Fibromodulin/FMOD in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Fibromodulin/FMOD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | Collagen-binding 59 kDa protein, fibromodulin, fibromodulin proteoglycan, FM, Keratan sulfate proteoglycan fibromodulin, SLRR2EKSPG fibromodulin |
| Host Species | Sheep |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human Fibromodulin/FMOD, Asp75-Ile376, Accession # Q06828 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?