Learn More
Invitrogen™ Fibroblasts Monoclonal Antibody (D7-FIB), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516642
Description
This antibody does not react with mouse or rat. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Fibroblasts/Epithelial Cells antibody, clone D7-FIB recognizes a approximately 112kDa molecule expressed on the cell surface of human fibroblasts.
Fibroblasts are the least specialized cells in the connective-tissue family. Thay are dispersed in connective tissue throughout the body, where they secrete a nonrigid extracellular matrix that is rich in type I and/or type III collagen. The activation of fibroblasts by inflammatory stimuli results in their migration, proliferation and deposition of extracellular matrix components, important features involved in both wound healing and fibrosis.
Specifications
| Fibroblasts | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| D7-FIB | |
| PE | |
| fibro-blasts | |
| Human foreskin fibroblasts. | |
| 100 Tests | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.