missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Antibody (ER-TR7), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64932PE
This item is not returnable.
View return policy
Description
Fibroblast Monoclonal specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Product Image.
Specifications
| Fibroblast | |
| Monoclonal | |
| PE | |
| reticular fibroblast | |
| Isolated C3H thymic stromal cells | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| ER-TR7 | |
| Flow Cytometry | |
| Rat | |
| Protein G purified | |
| RUO | |
| NB100-64932 recognizes ER-TR7, an antigen that is located in the cytoplasm of reticular fibroblasts and is a component of the extracellular matrix of lymphoid and non-lymphoid organs. The antigen recognized by clone ER-TR7 has not been identified but studies suggest that it is likely to be distinct from laminin, fibronectin, collagen types I-IV, heparin sulphate proteoglycan, entactin and nidogen. Clone ER-TR7 has been used to stain the microanatomy of various organs and also stains subendothelial deposits in atherosclerotic plaques. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction