missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Antibody (ER-TR7) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80736
This item is not returnable.
View return policy
Description
Fibroblast Monoclonal specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| Fibroblast | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry : 1:10-1:1000, Immunohistochemistry reported in scientific literature (PMID 22042977), Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunohistochemistry-Frozen : 1:500, CyTOF-ready | |
| reticular fibroblast | |
| Isolated C3H thymic stromal cells | |
| 0.125 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF | |
| ER-TR7 | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction