missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Mouse anti-Human, Clone: FAP/4851, Novus Biologicals™
Mouse Monoclonal Antibody
£219.00 - £423.00
Specifications
| Antigen | Fibroblast Activation Protein alpha/FAP |
|---|---|
| Clone | FAP/4851 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18349886
|
Novus Biologicals
NBP3-13883-20UG |
20 μg |
£219.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18393046
|
Novus Biologicals
NBP3-13883-100UG |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Fibroblast Activation Protein alpha/FAP | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Protein Phosphatase | |
| 10 mM PBS with 0.05% BSA | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| Recombinant fragment (around aa 1-200) of human Fibroblast Activation Protein alpha/FAP protein (exact sequence is proprietary) (Uniprot: Q12884) | |
| Primary | |
| Store at 4°C. |
| FAP/4851 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q12884 | |
| 2191 | |
| IgG2c κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title