missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Monoclonal Antibody (27C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120039
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-20039 detects fibrinogen in human samples.
Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.Specifications
Fibrinogen | |
Monoclonal | |
4.3 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P02671, P02675, P02679 | |
FGA, FGB, FGG | |
Fibrin degradation products. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
27C8 | |
Unconjugated | |
FGB | |
2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
Mouse | |
Protein A | |
RUO | |
2243, 2244, 2266 | |
4° C, do not freeze | |
Liquid |