missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma chain Antibody (OTI2D2), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70732G
This item is not returnable.
View return policy
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Fibrinogen gamma chain | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGG | |
| Full length recombinant protein of human Fibrinogen gamma chain (NP_000500) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2D2 | |
| Western Blot, Immunohistochemistry | |
| fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer | |
| 2266.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction