missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fibrinogen gamma chain Monoclonal antibody specifically detects Fibrinogen gamma chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, ELISA
Specifications
Specifications
| Antigen | Fibrinogen gamma chain |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1F2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH07044 |
| Gene Alias | fibrinogen gamma chain, fibrinogen, gamma polypeptide |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?