missing translation for 'onlineSavingsMsg'
Learn More

Fibrinogen beta chain Mouse anti-Human, Clone: 1F8E4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66186-1-IG-20UL

 View more versions of this product

Product Code. 16827835

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.
Specifications

Specifications

Fibrinogen beta chain
Monoclonal
1.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02675
FGB
Fibrinogen beta chain Fusion Protein Ag10205
20 μL
Primary
Human
Antibody
IgG2b
Western Blot
1F8E4
Unconjugated
FGB
FGB, fibrinogen beta chain
Mouse
Protein A
RUO
2244
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.