missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen beta chain Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-90956
This item is not returnable.
View return policy
Description
Fibrinogen beta chain Polyclonal specifically detects Fibrinogen beta chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Fibrinogen beta chain | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p, MGC104327, MGC120405 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human Fibrinogen beta chain antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FGB | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERKAPDAGGCLHADPDLGVLCPTGCQLQEALLQQERPIRNSVDELNNNVEAVSQTSSSSFQYMYLLKDLWQKRQKQVKDNENVVNEYSSELEKHQLYIDETVNSNIPTNLRVLRSILENLRSKIQKLE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2244 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction